Order Entry
Korea, Republic of
ContactUsLinkComponent
 
VWR®, Bottles, Wide Neck, Square, with Screw Cap
Catalog # VWRU414004-190
Supplier:  VWR Collection
undefined
VWR®, Bottles, Wide Neck, Square, with Screw Cap
Catalog # VWRU414004-190
Supplier:  VWR Collection

Specifications

  • Thread:
    100-415
  • Capacity:
    4000 ml
  • W×D×H:
    140×140×279 mm
  • Pk:
    1

Specifications

About this item

HDPE, translucent.

  • Features integrated handle for better grip
  • Space saving

Perfect for sample collection, storage and transport. Ideal for low temperature applications. The wide mouth allows easy filling and cleaning.