VWR®, Cryogenic protective aprons
Supplier: VWR Collection
Certificates
About this item
These aprons offer ideal body and leg protection against splashes and spills during the filling and handling of liquid nitrogen supply cylinders and bulk tanks, and during maintenance of cryogenic equipment.
- Ideal when filling liquid nitrogen supply cylinders and bulk tanks
- Adjustable straps with quick-release buckles provide added safety and convenience
Multiple layers of 100% nylon and 3M™ insulation allow for durable protection in low-temperature environments.
Specifications
- Catalog No:
- VWRU97008-290
- VWRU97008-292
- VWRU97008-294
- Garment application:
- Industrial
- Industrial
- Industrial
- Risk area:
- Cryo
- Cryo
- Cryo
- Gender:
- Unisex
- Unisex
- Unisex
- Fabric:
- 100% nylon
- 100% nylon
- 100% nylon