VWR®, Serological Pipettes, Disposable
Supplier: VWR Collection
Certificates
About this item
Borosilicate glass, Type I, Class B.
- Uniform pulled tip, calibrated to deliver
- Negative graduations increase versatility
- Non sterile, non plugged pipettes feature a blue scale
These disposable serological pipettes are available in sterile or non sterile formats, with a variety of packaging options to choose from. With two rings near the top denoting that the small amount remaining in the tip after free delivery has ceased must be blown out and added to the main volume delivered.
Packaging: Individually wrapped pipettes are packaged in colour-coded paper/plastic wraps and overpacked in convenient dispensing shelf packs for laboratory benches. Bulk pack pipettes are sealed in plastic bags and overpacked in shelf packs. Multi-pack pipettes come in colour-coded bags of 25 pipettes (1 and 2 ml sizes) or 10 pipettes (5 and 10 ml sizes).
Frequently Bought Together