VWR® Spinvane®, Magnetic stirring bars
Supplier: VWR Collection
Certificates
About this item
PTFE covered, Alnico V magnetic core. Spinvane® Triangle shaped magnetic stirring bars are designed for stirring in test tubes, centrifuge tubes and micro vials.
- The wings can also be trimmed to size if needed
- Autoclave safe
Frequently Bought Together