Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Volumetric Flasks, Class A, with Glass Stopper
VWR®, Volumetric Flasks, Class A, with Glass Stopper
  VWRU10546-066
VWR®, Volumetric Flasks, Class A, with Glass Stopper
  VWRU10546-066

 

  • Capacity:
    2 ml
  • Height:
    110 mm
  • Seq:
    01
  • Pk:
    6
  • Tolerance:
    ±0,015 ml

 

 

Manufactured from 3.3 expansion, low extractable borosilicate glass with standard taper glass stopper.

  • Calibrated to contain
  • With pennyhead glass stopper
  • Flasks feature a single graduation ring blasted on the neck and a marking spot

Sizes 5 ml and larger are designed from ASTM Specification E288, Class A requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.