
Sulphuric acid 95.0-98.0%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
: Avantor
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:14,08974
:∼335 °C (1013 hPa)
:3 °C
:1.84 g/cm³ (20 °C)
:Ambient
:231-639-5
:1830
:8,II
:GHS05
- For Laboratory,Research,or Manufacturing Use:95.0 - 98.0 %
- ACS - Assay (H₂SO₄):95.0 - 98.0 %
- Appearance:Passes Test
- ACS - Color (APHA):≤ 10
- ACS - Residue after Ignition:≤ 3 ppm
- ACS - Substances Reducing Permanganate (as SO₂):≤ 2 ppm
- Ammonium (NH₄):≤ 1 ppm
- Chloride (Cl):≤ 0.1 ppm
- Nitrate (NO₃):≤ 0.2 ppm
- Phosphate (PO₄):≤ 0.5 ppm
- Trace Impurities - Aluminum (Al):≤ 30.0 ppb
- Arsenic and Antimony (as As):≤ 4.0 ppb
- Trace Impurities - Barium (Ba):≤ 10.0 ppb
- Trace Impurities - Beryllium (Be):≤ 10.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 10.0 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 2.0 ppb
- Trace Impurities - Calcium (Ca):≤ 50.0 ppb
- Trace Impurities - Chromium (Cr):≤ 6.0 ppb
- Trace Impurities - Cobalt (Co):≤ 0.5 ppb
- Trace Impurities - Copper (Cu):≤ 1.0 ppb
- Trace Impurities - Gallium (Ga):≤ 10.0 ppb
- Trace Impurities - Germanium (Ge):≤ 10.0 ppb
- Trace Impurities - Gold (Au):≤ 10.0 ppb
- Heavy Metals (as Pb):≤ 500.0 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Lead (Pb):≤ 0.5 ppb
- Trace Impurities - Lithium (Li):≤ 10.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 7.0 ppb
- Trace Impurities - Manganese (Mn):≤ 1.0 ppb
- Trace Impurities - Mercury (Hg):≤ 0.5 ppb
- Trace Impurities - Molybdenum (Mo):≤ 10.0 ppb
- Trace Impurities - Nickel (Ni):≤ 2.0 ppb
- Trace Impurities - Niobium (Nb):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 500.0 ppb
- Trace Impurities - Selenium (Se):≤ 50.0 ppb
- Trace Impurities - Silicon (Si):≤ 100.0 ppb
- Trace Impurities - Silver (Ag):≤ 1.0 ppb
- Trace Impurities - Sodium (Na):≤ 500.0 ppb
- Trace Impurities - Strontium (Sr):≤ 5.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 10.0 ppb
- Trace Impurities - Thallium (Tl):≤ 20.0 ppb
- Trace Impurities - Tin (Sn):≤ 5.0 ppb
- Trace Impurities - Titanium (Ti):≤ 10.0 ppb
- Trace Impurities - Vanadium (V):≤ 10.0 ppb
- Trace Impurities - Zinc (Zn):≤ 5.0 ppb
- Trace Impurities - Zirconium (Zr):≤ 10.0 ppb
Frequently Bought Together