VWR®TraceClean®, Water sampling bottles, glass, wide neck
Type III (soda-lime) glass.
- Available with solid-top or open-top caps
- Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, PCBs, and phenolics analyses
- Open-top caps with ultrasonically bonded fluoropolymer resin/silicone septa are for use in volatile organic analysis
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers.' QA packers include a certificate of analysis in every case. QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Frequently Bought Together