Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Write-On Label Rolls, Magnetic
VWR®, Write-On Label Rolls, Magnetic
Catalog # VWRU80094-278
Supplier:  VWR Collection
undefined
VWR®, Write-On Label Rolls, Magnetic
Catalog # VWRU80094-278
Supplier:  VWR Collection
Specifications
  • Colour:
    White
  • Description:
    Write-On/wash-off labels
  • Dimensions:
    15,2 m×25 mm
  • Material:
    Vinyl
  • Pk:
    1
Specifications
About this item
This magnetic label material has a white vinyl surface that may be written on with wet-erase marker and then wiped off. Provided in a roll that may be cut to any size strip.
  • Thickness: 0,8 mm