VWR®, Absorbent mat pads and rolls
Supplier: VWR Collection
Certificates
About this item
Absorbent pads and rolls are ideal for spill clean-up; fast-wicking dimpled design absorbs liquids quickly to leave area clean and dry for improved safety. Efficient high-absorbency materials reduce waste of absorbents.
- Soak up leaks and spills fast, keeping floors at your workplace clean, dry and safe
- High-capacity absorbents stay on the job longer and are perfect for stocking spill kits
- Can be incinerated after use to reduce waste
These general purpose pads and rolls absorb water-based liquids, petroleum products and solvents for everyday maintenance and cleanup.
Frequently Bought Together