Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Filter Funnels according to Dr. Hirsch
VWR®, Filter Funnels according to Dr. Hirsch
VWR®, Filter Funnels according to Dr. Hirsch
VWR®, Filter Funnels according to Dr. Hirsch
Porcelain, glased inside and out, except rim.
Funnel with integral perforated filter plate can be used with paper or membrane discs.
Product Family Options
  • Length
    For filter Ø
    Funnel O-Ø
  • 70 mm
    13 - 15 mm
    42 mm
    1 * 1 items
    1 * 1 items - ₩66,887
    ₩66,887
    1 * 1 items
       ₩66,887
  • 87 mm
    25 - 30 mm
    50 mm
    1 * 1 items
    1 * 1 items - ₩78,985
    ₩78,985
    1 * 1 items
       ₩78,985
  • 132 mm
    40 - 47 mm
    78 mm
    1 * 1 items
    1 * 1 items - ₩108,685
    ₩108,685
    1 * 1 items
       ₩108,685