Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Büchner Funnels
VWR®, Büchner Funnels
Supplier:   VWR Collection
VWR®, Büchner Funnels
Supplier:  VWR Collection
Certificates
VWR®, Büchner Funnels
VWR®, Büchner Funnels
5 Options Available Below
Product Details & Documents
Porcelain, glased inside and out, except rim.
  • Resistant to chemicals
  • Very good temperature resistance
  • For paper or membrane filtration discs
Product Family Options
Product Information
  • Capacity
    For filter Ø
    Availability
    Price
  • 87 ml
    50 - 55 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩93,179
    ₩93,179
    1 * 1 items
       ₩93,179
  • 186 ml
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩122,467
    ₩122,467
    1 * 1 items
       ₩122,467
  • 550 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items
  • 700 ml
    110 - 124 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩284,892
    ₩284,892
    1 * 1 items
       ₩284,892
  • 1860 ml
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - ₩627,946
    ₩627,946
    1 * 1 items
       ₩627,946