VWR® VistaVision, Cover Glasses
Supplier: VWR Collection
Certificates
About this item
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.
- No disturbing self-fluorescence under UV light
- Suitable for use in coverslippers
Certifications: ISO 8255-1
Delivery: Rectangular cover glasses No. 1 are supplied 100 per hinged box. Rectangular cover glasses No. 1.5 are supplied 1 oz. per hinged box.
Frequently Bought Together