0
₩0
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
주문 입력
주문 상태
귀하의 계정
장바구니
Korea, Republic of
주문 입력
주문 상태
귀하의 계정
장바구니
ContactUsLinkComponent
Home
Racks
Tube Racks
VWR®, Racks for Microcentrifuge Tubes
VWR®, Racks for Microcentrifuge Tubes
VWR®, Racks for Microcentrifuge Tubes
VWRU20901-675
:
VWR Collection
VWR®, Racks for Microcentrifuge Tubes
VWRU20901-675
:
VWR Collection
For tubes:
1,5/2,0 ml
No. of Holes:
20
Description:
Rack for microcentrifuge tubes
W×D×H:
73×210×25 mm
Pk:
2
PP.
Easily transportable with one hand
Non-skid rubber feet
Autoclavable