Order Entry
Korea, Republic of
ContactUsLinkComponent
 
VWR®, Phenol-free total RNA purification kit
  N788-KIT
   Safer Human and Environmental Health
 :  
undefined
VWR®, Phenol-free total RNA purification kit
  N788-KIT
   Safer Human and Environmental Health
 :  

 

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Plant samples
    Bacteria
    Blood
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Phenol-Free Total RNA Purification Kit, 50 preps
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Description:
    Phenol-Free Total RNA Purification Kit

 

 

The phenol-free total RNA purification kit provides a rapid method for the isolation and purification of total RNA from cultured animal cells, tissue samples, blood, bacteria, yeast, fungi, plants and viruses.

  • Eliminate phenol and chloroform from RNA purification
  • Enhance recovery of total RNA, including microRNA species
  • Save time with a simple 20 minute protocol

The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). The RNA is preferentially purified from other cellular components, such as proteins.

The purified RNA, of the highest integrity can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.

 : Contains lysis solution, 40 ml; wash solution, 22 ml; elution solution, 6 ml; 50× mini spin columns; and 50× collection tubes.