Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Disposable Paper Funnel
VWR®, Disposable Paper Funnel
Disposable paper funnel, 186 ml
Catalog # 221-0274
   Sustainable Materials
Supplier:   VWR Collection
VWR®, Disposable Paper Funnel
Catalog # 221-0274
Supplier:  VWR Collection
   Sustainable Materials
VWR®, Disposable Paper Funnel
VWR®, Disposable Paper Funnel
Disposable paper funnel, 186 ml
Price
Inquire For Price
Log in for availability
1 * 25 items
Product Details & Documents
Made of clay-coated heavy paper stock.
  • Innovative 'backrest' allows hands-free use when pouring viscous liquids
  • Fits a wide variety of openings
  • Maximum flexibility - Cut stem to any opening size or shape
  • Apply a minimum of pressure and the funnel will stay in position
  • Use with virtually any liquid or powder*
Good alternative to disposable plastic funnels. Environmentally friendly paper product will naturally biodegrade in the standard municipal waste stream. Helps to reduce cross contamination.
Caution: * Not recommended for hot liquids.
Specifications
  • Capacity:
    186 ml
  • Description:
    Disposable paper funnel
  • Cat. No.:
    221-0274
  • Environmentally Preferable:
    Sustainable Materials