Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
₩0
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acetonitrile/Methanol/Water solvent kit
Acetonitrile/Methanol/Water solvent kit for HPLC
Acetonitrile/Methanol/Water solvent kit for HPLC
Acetonitrile/Methanol/Water solvent kit for HPLC
Catalog # 85047.001
Supplier:
VWR Chemicals
CAS Number:
Expand image
Acetonitrile/Methanol/Water solvent kit for HPLC
Catalog # 85047.001
Supplier:
VWR Chemicals
CAS Number:
Specifications
Pk:
1 SET
Seq:
0125
Specifications
About this item
Product Details
:
Product Details