

Potassium hydrogen sulphate, fused, powder, AR® ACS, Macron Fine Chemicals™
:7646-93-7
:
:KHSO₄
:MFCD00011404
:13,07695
:214 °C
:2.32 g/cm³ (20 °C)
:231-594-1
:2509
:8,II
:136.17 g/mol
:GHS05
:GHS07
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:37.5 - 38.6 %
- ACS - Acidity (as H₂SO₄):37.5 - 38.6 %
- ACS - Water (H₂O):≤2.5 %
- ACS - Chloride (Cl):≤0.002 %
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Heavy Metals (as Pb):≤0.001 %
- ACS - Iron (Fe):≤0.002 %
- ACS - Sodium (Na):≤0.01 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Ammonium Hydroxide Precipitate:≤0.01 %
- ACS - Calcium (Ca):≤0.002 %
- ACS - Magnesium (Mg):≤0.001 %
Frequently Bought Together