Choline chloride 98.0-100.5%, BAKER ANALYZED®, J.T.Baker®
Catalog # 1582-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:98.0 - 100.5 %
- Assay (anhydrous basis):98.0 - 100.5 %
- Residue after Ignition:≤0.10 %
- Ammonium Compounds:Passes Test
- Heavy Metals (as Pb):≤0.003 %
- Iron (Fe):≤0.002 %
Specification Test Results
About this item
:
:139.62 g/mol
Merck Index:13,02226
Melting Pt:244…247 °C
Density:∼1.205 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-655-4
CAS Number:67-48-1
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721