Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Filter Tips, Extra Long
VWR®, Filter Tips, Extra Long
UltraFine™ point, extra long, 10 µl, +, +, 10 racks, 96 each
Catalog # 732-1613
   Low Manufacturing Impact|Sustainable Packaging
Supplier:   VWR Collection
VWR®, Filter Tips, Extra Long
Catalog # 732-1613
Supplier:  VWR Collection
   Low Manufacturing Impact|Sustainable Packaging
VWR®, Filter Tips, Extra Long
VWR®, Filter Tips, Extra Long
UltraFine™ point, extra long, 10 µl, +, +, 10 racks, 96 each
Price
Inquire For Price
Log in for availability
1 * 960 items
Product Details & Documents
Aerosol filter tips with extended length.
  • Sustainable design based on virgin resins
  • Supplied sterile, autoclavable at 122 °C
  • RNase-/DNase-, human DNA-, endotoxin-, protease-, ATP- and BPA-free
Delivery information: Supplied in hinged racks, racks are refillable.
Specifications
  • CE-IVD:
    +
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    10 µl
  • Tip style:
    UltraFine™ point, extra long
  • Cat. No.:
    732-1613
  • Environmentally Preferable:
    Low Manufacturing Impact|Sustainable Packaging