

Copper(II) sulphate, anhydrous ≥99.0%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
:GHS07
:GHS09
Specification Test Results
- Assay (calculated on dried substance):Min. 99.0 %
- Identification:Passes test
- Insolubility in water:Max. 100 ppm
- Loss on drying (250°C):Max. 1 %
- Substances not precipitated by H2S:Max. 0.2 %
- Total N (Nitrogen):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- Fe (Iron):Max. 50 ppm
- Pb (Lead):Max. 100 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number84845_COPPER (II) SULPHATE ANHYDROUS_BSE_REV0
BSE/TSE Certificates
spec_VWRC84845.230 spec_VWRC84845.230 spec_VWRC84845.290 spec_VWRC84845.290
Product specification
Frequently Bought Together