Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
₩0
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antimony trichloride solutions
Antimony trichloride solution R1 Reag. Ph. Eur. 1007702
Antimony trichloride solution R1 Reag. Ph. Eur. 1007702
Antimony trichloride solution R1 Reag. Ph. Eur. 1007702
Catalog # 87780.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Antimony trichloride solution R1 Reag. Ph. Eur. 1007702
Catalog # 87780.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
100 mL
Seq:
2440
Specifications
About this item
Product Details
Documentation
:
EINECS:
0
Product Details
Documentation