

Zinc oxide ≥99.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1314-13-2
:
Formula:ZnO
MDL Number:MFCD00011300
:81.39 g/mol
Merck Index:13,10200
Melting Pt:1975 °C
Density:5,61 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-222-5
UN:3077
ADR:9,III
REACH:01-2119463881-32
:GHS09
:GHS07
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤0.01%
- ACS - Assay (ZnO):≥99.0 %
- ACS - Insoluble in Dilute H₂SO₄:≤0.01 %
- ACS - Alkalinity:Passes Test
- ACS - Nitrate (NO₃):≤0.003 %
- ACS - Sulfur Compounds (as SO₄):≤0.005 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Lead (Pb):≤0.005 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.05 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Manganese (Mn):≤5.0 ppm
Specification Test Results
Frequently Bought Together