Aluminium oxide ≥98.5%, powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 0536-05
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay (Al₂O₃):≥ 98.5 %
- Loss on Ignition:≤ 1.0 %
- Chloride (Cl):≤ 0.005 %
- Sulfate (SO₄):≤ 0.005 %
- Heavy Metals (as Pb):≤ 0.001 %
- Iron (Fe):≤ 0.02 %
Specification Test Results
About this item
:
:101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3.94 g/cm³ (20 °C)
EINECS:215-691-6
REACH:01-2119529248-35
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424