Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
CEDuoCleanTechDuoSponge-955x690.jpg
CE Duo® CleanTech Duo® Sponge
Microfiber cleaning and disinfecting for hard to reach areas! The CE Duo CleanTech Sponge allows you to disinfect while cleaning hard-to-reach areas in cleanrooms and controlled environments. The patented microfiber composition removes contamination from surfaces and holds it within the sponge.
Learn more
CE Duo® CleanTech Duo® Sponge | Avantor
The microfiber laminated foam is ideal for cleaning and disinfecting all types of surfaces and hard-to-reach areas, such as behind equipment, cabinets and piping, as well as hoods, isolators, door handles, transition surfaces and all other surfaces where a mop cannot reach. The sponge can be used with either the bucket or pre-prepared method. The double sided, hand-held design results in a higher degree of user control.