Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
cell-culture.jpg
From Culture to Discovery
Researchers all over the world choose Nunc microplates for their high quality, consistency and variety of available formats.
Thermo Scientific Cell Culture Plastics | Avantor
current-promotion.jpg
  • Thermo Scientific Nunc 15 mL and 50 mL Conical Centrifuge Tubes - Buy 2 cases and receive one case of Serological Pipettes at no additional charge!
See all Thermo Scientific Promotions
Well-Plates.jpg
Featured Product
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo ScientificNunc Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays.
Learn More
Product selection
cell-dishes-540.jpg
Dishes in more than 100 combinations of format and surface. Excellent optical quality for manual or automated imaging
cell-flask-540.jpg
Prepare and store culture media and perform a broad range of culturing techniques with our diverse selection of flat-sided flasks and accessories, plastic spinner flasks and plastic Erlenmeyer flasks
microplates-540.jpg
Researchers all over the world choose Nunc microplates for their high quality, consistency and variety of available formats
cell-culture-tube-540.jpg
Researchers all over the world choose Nunc microplates for their high quality, consistency and variety of available formats
conical-tubes-540.jpg
Take advantage of the improved separation and pelleting of a conical tube while eliminating risk of breakage with our polycarbonate and polystyrene tubes