Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
honeywell-955.jpg
Original and Accurate Hydranal™ Karl Fischer Reagents
Original and Accurate Hydranal™ Karl Fischer Reagents​ | Avantor
Leading the way in innovation and safety since 1979, Hydranal™ reagents for Karl Fischer (KF) titration have been recognized as the most trusted brand in the industry.
The Hydranal portfolio is made up of methanol and ethanol-based KF reagents free from pyridine. Reagents for volumetric and coulometric application are completed by a wide range of standards and auxiliaries.
Hydranal products are known for their high quality and consistency, and are backed by dual accreditation, ISO/IEC 17025 and ISO 17034.
More than 700 Hydranal lab reports can be downloaded from www.lab-honeywell.com/hydranal-search/
Product selection
24615996-540x390.jpg
Volumetric Karl Fischer Titration
Explore the broadest portfolio of one- and two-component volumetric reagents and a selection of special solvents for volumetric titrations.
24616000-540x390.jpg
Coulometric Karl Fischer Titration
Coulometric reagents based on different solvent compositions serve to support the broad variety of samples analyzed by Karl Fisher titration.
24616003-540x390.jpg
Exactly confirmed water content for titer determination, for precision & accuracy analysis and validation & control of Karl Fischer titrators.
24616002-540x390.jpg
Additional Karl Fischer titration products, incl. solubilizers, buffers to neutralize acidic or basic samples, drying agents and a Moisture Test Kit.