Order Entry
Korea, Republic of
ContactUsLinkComponent
Trio.bas955x590.jpg
TRIO.BAS
The most complete line of impact microbial air sampling instruments available
TRIO.BAS Microbial Air Samplers are available in Petri, Contact plate, or Compact Dry formats, 100-200L per minute models, and a variety of ergonomic, lightweight platforms that make active impact air monitoring a breeze. Battery life of 15 hours on a single charge and no plugs or external connections reduce the risk of contamination.  Always expanding, the TRIO.BAS line offers the most utility of any air sampler with the addition of the new AIRBIO DUO, RABS, and Multiflex models.
TRIO.BAS, the future of air sampling here today!
triocalibrationservices-260x190.jpg
TRIO.BAS Calibration Services
To better service our customers, Hardy Diagnostics offers complete calibration and repair services for TRIO BAS Air Samplers.
Repair Service Form
Product selection
Trio.BasMONO-Kits400x495.jpg
Trio.Bas™ MONO Kit
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles. The Trio.Bas MONO Kit features one aspirating head.
TrioBasDUOKits400x495.jpg
Trio.Bas™ DUO Kit
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles. The Trio.Bas DUO Kit features dual aspirating heads.
triobastrio400x495.jpg
Trio.Bas™ TRIO Kit
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.The Trio.Bas TRIO Kit features three aspirating heads.
Trio-gas-kit400x495.jpg
Trio.Bas™ Gas Kit
The Trio.Bas GAS system is designed to test the presence of microorganisms in compressed gas samples supplied from tanks and pipes under pressure.