Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
AzureSapphire955x590.jpg
Azure Sapphire™ Biomolecular Imager
The next generation lab imaging platform. This system supports long and short wavelengths of near infrared fluorescence (NIR), red/green/blue (RGB) imaging, chemiluminescent imaging, phosphor imaging as well as optical densitometry (OD) of proteins in stained gels.
Azure Sapphire™ Biomolecular Imager | Avantor
Product Features
  • Up to four solid state lasers (488, 520, 658 and 784 nm) offering ultimate excitation sensitivity
  • Photomultiplier tube (PMT) for fluorescence and phosphor imaging, avalanche photodiodes (APD) for near-infrared imaging and a CCD sensor for chemiluminescent and visible imaging
  • Ultra-wide dynamic range for imaging and quantifying low and high abundance samples simultaneously
  • Image resolution down to 10 microns for high-quality image analysis
  • Full integration with Sapphire Capture and AzureSpot software programs for perfect imaging and accurate analysis every time
BreakthroughImage_400x495.jpg
Breakthrough image capture and analysis
Exceptional data quality through extremely sensitive detection, ultra high resolution and broad linear dynamic range
Learn more