JERRICAN HDPE 10L SQUARE UN APP CAP STERILE 1 * 6 items
Catalog # 331-0415
Supplier: VWR Collection
New Product
JERRICAN HDPE 10L SQUARE UN APP CAP STERILE 1 * 6 items
Catalog # 331-0415
Supplier: VWR Collection
New Product
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, フェイシャルティッシュ、
Supplier: VWR Collection
VWR®, ニトリルグローブ
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, ニトリルグローブ、ニトリルライト
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals