Orders LinkContactUsLinkComponent
J.T.Baker® プレート SRW、マグネティック
Tip comb with barcode, Transparent (Removed), -
Catalog # 43001-0515
   Low Manufacturing Impact
Supplier:   Avantor
J.T.Baker® プレート SRW、マグネティック
Catalog # 43001-0515
Supplier:  Avantor
   Low Manufacturing Impact
J.T.Baker® プレート SRW、マグネティック
J.T.Baker® プレート SRW、マグネティック
Tip comb with barcode, Transparent (Removed), -
Price
$738.38
Log in for availability
1 * 60 items
Product Details & Documents
These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as the KingFisher™ Apex, Flex 96 and Presto.
  • Standard SBS footprint
  • Cylindrical wells enable good sample mixing
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code assists registration and identification of individual samples
Specifications
  • Sterile:
    -
  • Colour:
    Transparent (Removed)
  • 説明:
    Tip comb with barcode
  • Cat. No.:
    43001-0515
  • Environmental certification:
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
  • Environmentally Preferable:
    Low Manufacturing Impact