Japan
ContactUsLinkComponent
VWR Filter Combination, 50mm , PTFE , 0.22 μm filter , 3/8 inch ( 9.5 mm ) inner diameter hose , large mouth conversion cover ,Sterile,1 set/bag,1 bag/box 1 * 1 items
Catalog # VWRA734-3674
Supplier:  VWR Collection
New Product
VWR Filter Combination, 50mm , PTFE , 0.22 μm filter , 3/8 inch ( 9.5 mm ) inner diameter hose , large mouth conversion cover ,Sterile,1 set/bag,1 bag/box 1 * 1 items
Catalog # VWRA734-3674
Supplier:  VWR Collection
New Product
Frequently Bought Together