ContactUsLinkComponent
PFAS KIT 2IN1 548-0029A+548-7012A 1.5ml Short Thread Vial, ND9, 32 x 11.6mm, clear glass, 1st hydrolytic class, wide opening, label and filling lines + 9mm PFAS PP Short Thread Cap ND9, light-green, with hole Silicone white/Polypropylene transparent 1 * 1 items
Catalog # VWRI548-7014A
Supplier:  VWR Collection
New Product
PFAS KIT 2IN1 548-0029A+548-7012A 1.5ml Short Thread Vial, ND9, 32 x 11.6mm, clear glass, 1st hydrolytic class, wide opening, label and filling lines + 9mm PFAS PP Short Thread Cap ND9, light-green, with hole Silicone white/Polypropylene transparent 1 * 1 items
Catalog # VWRI548-7014A
Supplier:  VWR Collection
New Product