Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HDPE AMB BOTL RECT 2700ML CAP YEL ST 1 * 30 items
Catalog # 215-4652
Supplier:
VWR Collection
Certificates
Expand image
HDPE AMB BOTL RECT 2700ML CAP YEL ST 1 * 30 items
Catalog # 215-4652
Supplier:
VWR Collection
Certificates
Related Information
Documentation
Datasheet
TDS HDPE Bottle.pdf
VWRI215-4652_2311260887.pdf
Documentation
Frequently Bought Together
VWR®, VWR® Standard Line, Transfer pipettes
Supplier: VWR Collection
VWR®, ニトリルグローブ
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
BOTTLE PET 250ML ROUND CAP WHITE 1 * 150 items
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals