Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 防振秤量面

VWR®, 防振秤量面

Anti-vibration weighing surface, 450x300x40 mm, 20 kg
Catalog # 611-5275
Supplier:   VWR Collection
VWR®, 防振秤量面
Catalog # 611-5275
Supplier:  VWR Collection
VWR®, 防振秤量面
VWR®, 防振秤量面

Anti-vibration weighing surface, 450x300x40 mm, 20 kg

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Polished granite. Four absorbing feet dampen the transmission of extraneous vibrations to this slab making it suitable, not only for high precision weighing, but also microscopy and other similar delicate instrumentation.

Weight: 44 kg

Documents

Specifications
  • 説明:
    Anti-vibration weighing surface
  • W×D×H:
    450x300x40 mm
  • Weight:
    20 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.