Orders LinkContactUsLinkComponent
VWR®, クリーンルーム用オーバーオール
VWR®, クリーンルーム用オーバーオール
White, 4XL, +
Catalog # VWRA114-9330
Supplier:   VWR Collection
VWR®, クリーンルーム用オーバーオール
Catalog # VWRA114-9330
Supplier:  VWR Collection
VWR®, クリーンルーム用オーバーオール
VWR®, クリーンルーム用オーバーオール
White, 4XL, +
Price
Inquire For Price
Log in for availability
1 * 20 items
Product Details & Documents
These sterile, single-use overalls are made of breathable SMS fabric. They are ideal for use in cleanroom production and research.
  • Hood and two-way closure
  • Elasticated wrist, ankles and back for tighter fit and easier donning
  • Thumb loops (latex-free)
  • Bound seams
  • Without boots
  • Folded with snap release to arms and legs to aid aseptic donning
Suitable for use in ISO Class 5 + 6 cleanroom environments.
Weight: 60 g
Level I acc. IEST-RP-CC003.3
Certifications: Type 5/6; Cat. III; EN 1149-5.
包装: Individually double-packed in easy-tear bags. 20 overalls per liner.
Specifications
  • 衣服のサイズ:
    4XL
  • Sterile:
    +
  • Colour:
    White
  • Seq:
    07
  • Cat. No.:
    VWRA114-9330
  • Cleanroom class:
    100/ISO 5
  • Model:
    With hood
  • Garment application:
    Cleanroom|Laboratory
  • Weight:
    60 g