Reagent Bottle with cap, Narrow mouth, LDPE, 8ml 1 * 12 items
Catalog # VWRA215-4601
Supplier: VWR Collection
Reagent Bottle with cap, Narrow mouth, LDPE, 8ml 1 * 12 items
Catalog # VWRA215-4601
Supplier: VWR Collection
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, ニトリルグローブ
Supplier: VWR Collection
VWR®, フェイシャルティッシュ、
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, ニトリルグローブ、ニトリルライト
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection