Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
Orange veiwing glasses for use with blue LED illuminators, allows clear viewing of fluorescing DNA bands, includes case 1 * 1 items
700-1488
:
VWR Collection
Orange veiwing glasses for use with blue LED illuminators, allows clear viewing of fluorescing DNA bands, includes case 1 * 1 items
700-1488
:
VWR Collection
Frequently Bought Together
VWR® VisiCam® Plus, Digital USB Cameras
: VWR Collection