Masterflex® Cleanroom 1" Sanitary Flange × ½" Hose Barb in PVDF; 10 pack
MFLX40330-39
Maintain sanitary conditions with secure connections from sanitary Tri-Clamp to tubing.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
- Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in the biopharmaceutical, food and beverage, and single-use industries. They have excellent chemical resistance and may be sterilized by gamma irradiation, EtO, E-beam, or autoclave. The PVDF fittings withstand higher temperatures than the Polypropylene fittings. Clamps and gaskets required for a complete system are sold separately.
- バーブサイズ/チューブ内径 (ポート2):½"
- 接続タイプ:Sanitary to Barb
- Flow pattern:Straight
- Sanitary clamp size (Port 1):1½"
- 説明:Cleanroom 1½" sanitary tri-clamp × ½" barb adapter in PVDF; 10 pack
- Seq:07
- Environmentally Preferable:
- No. of ports:2
- Regulatory approvals:FDA;USP
- Sterilisation:Gamma irradiation, EtO, autoclave
- Flange size:1"
- Pk:10
- Fitting material:PVDF
- Sanitary gasket size:1½"
Frequently Bought Together