ContactUsLinkComponent
VWR Polypropylene Film, Non-Sterile. Ideal for short-term storage, incubation/ELISA. Temperature Range: -20°C to +120°C. Secure seal minimizes evaporation and reduces aerosols upon removal. 1 * 100 items
  391-0677
VWR Polypropylene Film, Non-Sterile. Ideal for short-term storage, incubation/ELISA. Temperature Range: -20°C to +120°C. Secure seal minimizes evaporation and reduces aerosols upon removal. 1 * 100 items
  391-0677
Frequently Bought Together