Masterflex® Fitting, Natural Polypropylene, Elbow, Hose Barb Union, 1/8" ID; 100/PK
Catalog # MFLX40626-71
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/8" ID
Create 90° bends in flexible tubing—without the kinks!
- バーブサイズ/チューブ内径 (ポート1):⅛"
- バーブサイズ/チューブ内径 (ポート2):⅛"
- 接続タイプ:Barbed Union
- Flow pattern:Elbow
- 説明:Fitting, Natural Polypropylene, Elbow, Hose Barb Union, ⅛" ID; 100/PK
- Seq:21
- Environmentally Preferable:
- Model:40626-71
- Sterilisation:Ethylene Oxide (ETO)
- Pk:100
- Fitting material:Polypropylene
Frequently Bought Together