Masterflex® Replacement Tubing Set, Norprene®, 0.02 to 54 GPH Flow Range
Catalog # MFLX74204-44
Supplier: Avantor Fluid Handling
About this item
Choose tubing based on chemical compatibility, flow rate, and your current pump.
- Suitable for use with your Masterflex® High-Pressure Peristaltic Pump
Choose the tubing that is compatible with your pump and meets your application needs.
- 最大流量:7000 ml/min
- Max. pressure:8,5 bar (125 psi)
- Max. temperature:85 °C (185 °F)
- 説明:Replacement Tubing Set, Norprene®, 0,02 to 54 gal./h Flow Range
- Seq:11
- Environmentally Preferable:
- Model:74204-44
- Wetted materials:LLDPE, Norprene, PVC, PVDF
- Inlet connection:1/2" hose barb
- Outlet connection:1/2" hose barb
- Suction lift, wet:9144 mm of H₂O (360" H₂O)
- Max. viscosity:12000 cP
- Min. flow rate:1,4 ml/min
- Warranty:2 years
- Pk:1
- Max. motor RPM:125 rpm
Frequently Bought Together