Masterflex® Fitting, 316 Stainless Steel, Straight, Female Thread Reducer, 1/8" x 1/2" NPT(F)
Catalog # MFLX31410-13
Supplier: Avantor Fluid Handling
About this item
Maintain a permanent connection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.
- 接続タイプ:Threaded Reducer
- Flow pattern:Straight
- Thread size NPT(F) (port 1):½"
- Thread size NPT(F) (port 2):⅛"
- 説明:Fitting, 316 Stainless Steel, Straight, Female Thread Reducer, ⅛" × ½" NPT(F)
- Seq:09
- Environmentally Preferable:
- Model:31410-13
- Fitting depth:8,73 mm
- Sterilisation:Autoclave, ethylene oxide (ETO), or gamma
- Warranty:1 year
- Pk:1
- Fitting material:Stainless Steel 316
Frequently Bought Together