Masterflex® Fitting, Polypropylene, Elbow, Hose Barb Union, 1/16" ID; 10/PK
Catalog # MFLX51518-49
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/16" ID
Create 90° bends in flexible tubing—without the kinks!
- バーブサイズ/チューブ内径 (ポート1):¹⁄₁₆"
- バーブサイズ/チューブ内径 (ポート2):¹⁄₁₆"
- 接続タイプ:Barbed Union
- Flow pattern:Elbow
- 説明:Fitting, Polypropylene, Elbow, Hose Barb Union, ¹⁄₁₆" ID; 10/PK
- Seq:34
- Environmentally Preferable:
- Model:51518-49
- Pk:10
- Fitting material:Polypropylene
Frequently Bought Together