Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Thread, 7/16" Hex x 1/8-27; 25/PK
Catalog # MFLX45518-86
Supplier: Avantor Fluid Handling
CE certificate
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Connection Type: Male Luer Lock to Threaded, 7/16" Hex x 1/8-27
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- 接続タイプ:Male Luer to Thread
- Flow pattern:Straight
- 説明:Fitting, Polypropylene, Straight, Male Luer Lock to Thread, ⁷⁄₁₆" Hex x ⅛-27; 25/PK
- Seq:09
- Environmentally Preferable:
- Model:45518-86
- Connection type (port 1):Male luer lock
- Connection type (port 2):⅛-27 NPT thread
- No. of ports:2
- Pk:25
- Fitting material:Polypropylene
- Thread size UNF(M) (port 1):⅛"-27
Frequently Bought Together