Masterflex® Flowmeter Tripod Base Stand for 32461-Series Flowmeters
Catalog # MFLX32462-55
Supplier: Avantor Fluid Handling
About this item
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
- 説明:Tripod base Flowmeter Stand For 32461
- Seq:07
- Environmentally Preferable:
- Model:32462-55
- Pk:1
Frequently Bought Together