ContactUsLinkComponent
Masterflex® Flowmeter Tripod Base Stand for 32461-Series Flowmeters
Masterflex® Flowmeter Tripod Base Stand for 32461-Series Flowmeters
Catalog # MFLX32462-55
undefined
Masterflex® Flowmeter Tripod Base Stand for 32461-Series Flowmeters
Catalog # MFLX32462-55
About this item
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
  • 説明:
    Tripod base Flowmeter Stand For 32461
  • Seq:
    07
  • Environmentally Preferable:
  • Model:
    32462-55
  • Pk:
    1
Frequently Bought Together