Masterflex® Fitting, Nylon, Y-Connector, Hosebarb Reducer, 1/8" ID x 1/8" ID x 1/4" ID; 10/PK
Catalog # MFLX30726-31
Supplier: Avantor Fluid Handling
About this item
Ensure a tight seal every time.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- バーブサイズ/チューブ内径 (ポート1):1/8"
- バーブサイズ/チューブ内径 (ポート2):1/8"
- 接続タイプ:Barbed Reducer
- Flow pattern:Y
- 説明:Fitting, Nylon, Y-Connector, Hosebarb Reducer, 1/8" ID x 1/8" ID x 1/4" ID; 10/PK
- Seq:07
- Environmentally Preferable:
- Model:30726-31
- Sterilisation:Ethylene oxide (ETO) only
- Pk:10
- Barb size/tubing int.Ø (port 3):1/4"
- Fitting material:Nylon
Frequently Bought Together