Masterflex® Remote Control Cable Connectors, Avantor®
DB9 male 9-pin connector for use with drives
Catalog # MFLX07595-45
Supplier: Avantor Fluid Handling
Masterflex® Remote Control Cable Connectors, Avantor®
Catalog # MFLX07595-45
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Create your own cable for optimization of your Masterflex® drive.
- Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
Documents
Masterflex LS MasterSense Drives Pump
Product Literature
Specifications
- 説明:DB9 male 9-pin connector for use with drives
- Cat. No.:MFLX07595-45
- Model:07595-45