Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Multichannel Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty, Stainless steel, Masterflex L/S® Multichannel Pump Head, L/S
Catalog # MFLX07534-01
Supplier: Avantor Fluid Handling
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Catalog # MFLX07534-01
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Documents
Certificates
Search by Lot NumberSpecifications
- 説明:L/S® Multichannel Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty
- Material:Stainless steel
- Seq:06
- Compatible with:Masterflex L/S® Multichannel Pump Head
- Pump series:L/S