Masterflex® I/P® Tubing Loading Key for Standard Pump Heads
Catalog # MFLX07019-90
Supplier: Avantor Fluid Handling
About this item
Ensure proper tubing alignment and installation.
- Constructed of durable stainless steel
L/S® and I/P® Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S® or I/P®.
- 説明:tubing loading key for I/P Standard pump heads.
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
- Pump series:I/P
Frequently Bought Together