Masterflex® Fitting, Antimicrobial Polypropylene, Straight, Hose Barb Union, 1/4" ID; 10/PK
Catalog # MFLX06368-04
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Chemical resistant
- Connection Type: 1/4" ID
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- バーブサイズ/チューブ内径 (ポート1):¼"
- バーブサイズ/チューブ内径 (ポート2):¼"
- 接続タイプ:Barbed Union
- Flow pattern:Straight
- 説明:Fitting, Antimicrobial Polypropylene, Straight, Hose Barb Union, ¼" ID; 10/PK
- Seq:11
- Environmentally Preferable:
- Model:06368-04
- Orifice dimensions:3,56 mm (0,140625")
- Sterilisation:Ethylene oxide (ETO) or gamma
- Pk:10
- Fitting material:Polypropylene
Frequently Bought Together